Learn More
Selleck Chemical LLC Amlodipine Besylate 50mg 111470-99-6
SDP
Supplier:  Selleck Chemical LLC S181350MG
Encompass_Preferred
Amlodipine Besylate is a long-acting calcium channel blocker, used to lower blood pressure and prevent chest pain. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-540-5