Learn More
Medchemexpress LLC Amlodipine Besylate, USP | 111470-99-6 | MFCD00887594 | >=98% | 1 G
SDP
Catalog No. 5000406362
Encompass_Preferred
Amlodipine besylate is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, which inhibits the initial influx of calcium. This compound is suitable for research related to high blood pressure and cancer.
  • Antianginal agent: Used in research for angina.
  • Orally active: Convenient for oral administration in research settings.
  • Dihydropyridine calcium channel blocker: A specific type of calcium channel blocker with a well-defined mechanism of action.
  • Mechanism of action: Blocks voltage-dependent L-type calcium channels, inhibiting calcium influx.
  • Versatile research applications: Applicable for research concerning high blood pressure and cancer.
Catalog No. 50-004-06362 Supplier Medchemexpress LLC Supplier No. HYB0317B1G
Amlodipine besylate is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, which inhibits the initial influx of calcium. This compound is suitable for research related to high blood pressure and cancer.
  • Antianginal agent: Used in research for angina.
  • Orally active: Convenient for oral administration in research settings.
  • Dihydropyridine calcium channel blocker: A specific type of calcium channel blocker with a well-defined mechanism of action.
  • Mechanism of action: Blocks voltage-dependent L-type calcium channels, inhibiting calcium influx.
  • Versatile research applications: Applicable for research concerning high blood pressure and cancer.