Learn More
Medchemexpress LLC Amlodipine-d4 | 1185246-14-3 | 98.5% | 5 MG
SDP
Catalog No. 5000406884
Encompass_Preferred
Amlodipine-d4 is a deuterium labeled Amlodipine. Amlodipine is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, which inhibits the initial influx of calcium. Amlodipine can be utilized for the research of high blood pressure and cancer.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06884 Supplier Medchemexpress LLC Supplier No. HYB0317S15MG
Add to Cart
Add to Cart
Amlodipine-d4 is a deuterium labeled Amlodipine. Amlodipine is an antianginal agent and an orally active dihydropyridine calcium channel blocker. It functions by blocking voltage-dependent L-type calcium channels, which inhibits the initial influx of calcium. Amlodipine can be utilized for the research of high blood pressure and cancer.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS