Learn More
Medchemexpress LLC Amlodipine-d4 (maleate) | 2714486-25-4 | 98.2% | 5 MG
SDP
Catalog No. 5000406700
Encompass_Preferred
Amlodipine-d4 (maleate) is the deuterium-labeled Amlodipine maleate, a dihydropyridine calcium channel blocker. It functions as an orally active antianginal agent by blocking voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. This compound is valuable for research into high blood pressure and cancer.
  • Functions as a tracer in drug development processes.
  • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Aids in the study of high blood pressure and cancer.
  • Offers potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06700 Supplier Medchemexpress LLC Supplier No. HYB0317AS5MG
Amlodipine-d4 (maleate) is the deuterium-labeled Amlodipine maleate, a dihydropyridine calcium channel blocker. It functions as an orally active antianginal agent by blocking voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium. This compound is valuable for research into high blood pressure and cancer.
  • Functions as a tracer in drug development processes.
  • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Aids in the study of high blood pressure and cancer.
  • Offers potential to influence the pharmacokinetic and metabolic profiles of drugs.