Learn More
Apexbio Technology LLC AMLODIPINE-MALEATE-500MG
SDP
Catalog No. 5000569344
Encompass_Preferred
Amlodipine maleate (CAS No 88150-47-4) is a calcium channel blocker commonly used in cardiovascular research Originating from the dihydropyridine class its mechanism of action involves inhibiting the influx of calcium ions through L-type calcium channels leading to vasodilation and reduced blood pressure This compound has demonstrated in vitro activity in the nanomolar to low micromolar range making it a valuable tool for studying cardiac function and vascular smooth muscle relaxation It is often utilized in cell models including cardiomyocytes and endothelial cells as well as in animal studies to explore hypertension and related pathologies Additionally amlodipine maleate is employed in high-throughput drug screening assays to identify potential therapeutic agents Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69344 Supplier Apexbio Technology LLC Supplier No. C6705500
Amlodipine maleate (CAS No 88150-47-4) is a calcium channel blocker commonly used in cardiovascular research Originating from the dihydropyridine class its mechanism of action involves inhibiting the influx of calcium ions through L-type calcium channels leading to vasodilation and reduced blood pressure This compound has demonstrated in vitro activity in the nanomolar to low micromolar range making it a valuable tool for studying cardiac function and vascular smooth muscle relaxation It is often utilized in cell models including cardiomyocytes and endothelial cells as well as in animal studies to explore hypertension and related pathologies Additionally amlodipine maleate is employed in high-throughput drug screening assays to identify potential therapeutic agents Experimental concentrations typically depend on the specific research design