Learn More
Medchemexpress LLC Amlodipine maleate | 88150-47-4 | 99.9% | 1 G
SDP
Catalog No. 5000406672
Encompass_Preferred
Amlodipine maleate is a dihydropyridine calcium channel blocker that acts as an orally active antianginal agent. It blocks voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium. It can be used for the research of high blood pressure and cancer.
  • Calcium channel blocker
  • Orally active
  • Blocks voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium
  • Used for research related to high blood pressure and cancer
  • High purity (99.87%)
Catalog No. 50-004-06672 Supplier Medchemexpress LLC Supplier No. HYB0317A1G
Amlodipine maleate is a dihydropyridine calcium channel blocker that acts as an orally active antianginal agent. It blocks voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium. It can be used for the research of high blood pressure and cancer.
  • Calcium channel blocker
  • Orally active
  • Blocks voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium
  • Used for research related to high blood pressure and cancer
  • High purity (99.87%)