Learn More
Medchemexpress LLC Amlodipine maleate | 88150-47-4 | 99.9% | 1 ML
SDP
Catalog No. 5000406594
Encompass_Preferred
Amlodipine maleate is a dihydropyridine calcium channel blocker that acts as an orally active antianginal agent. It blocks voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium.
  • Acts as an orally active antianginal agent
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits the initial influx of calcium
  • Used for research on high blood pressure
  • Used for research on cancer
Catalog No. 50-004-06594 Supplier Medchemexpress LLC Supplier No. HYB0317A10MM/1ML
Amlodipine maleate is a dihydropyridine calcium channel blocker that acts as an orally active antianginal agent. It blocks voltage-dependent L-type calcium channels, thereby inhibiting the initial influx of calcium.
  • Acts as an orally active antianginal agent
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits the initial influx of calcium
  • Used for research on high blood pressure
  • Used for research on cancer