Learn More
Medchemexpress LLC Amlodipine maleate | 88150-47-4 | 99.9% | 5 G
SDP
Catalog No. 5000406361
Encompass_Preferred
Amlodipine maleate is a dihydropyridine calcium channel blocker that acts as an orally active antianginal agent. It blocks voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium. This compound is used in research for high blood pressure and cancer studies.
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits initial influx of calcium
  • Attenuates uridine 5′-triphosphate (UTP)-induced increases of intracellular calcium
  • Inhibits store-operated calcium influx
Catalog No. 50-004-06361 Supplier Medchemexpress LLC Supplier No. HYB0317A5G
Amlodipine maleate is a dihydropyridine calcium channel blocker that acts as an orally active antianginal agent. It blocks voltage-dependent L-type calcium channels, inhibiting the initial influx of calcium. This compound is used in research for high blood pressure and cancer studies.
  • Blocks voltage-dependent L-type calcium channels
  • Inhibits initial influx of calcium
  • Attenuates uridine 5′-triphosphate (UTP)-induced increases of intracellular calcium
  • Inhibits store-operated calcium influx