Learn More
Aqua Solutions Ammonia Nitrogen Standard APHA 10.0 ppm as NH₃ (1ml = 0.01 mg NH₃) (4L)
SDP
Catalog No. NC9462357
Encompass_Preferred
Ammonia Nitrogen Standard APHA 10.0 ppm as NH₃ (1ml = 0.01 mg NH₃) (4L)
Catalog No. NC9462357 Supplier Aqua Solutions Supplier No. CMS001
Ammonia Nitrogen Standard APHA 10.0 ppm as NH₃ (1ml = 0.01 mg NH₃) (4L)