Learn More
STEMCELL Technologies Ammonium Chloride Solution, Size: 100 mL
SDP
Catalog No. NC9041684
Encompass
Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells. It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes. It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis.
Catalog No. NC9041684 Supplier STEMCELL Technologies Supplier No. 07800
Ammonium Chloride Solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen, or bone marrow cells. It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes. It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis.