Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Ammonium Hydroxide 6% V/V, 4l

Catalog No. 504400775
Encompass_Preferred

Ammonium Hydroxide 6% V/V, 1336-21-6, WT%: <10%, 4l

Catalog No. 50-440-0775 Supplier LaMarKa Chemical Company Supplier No. 00006050660132939
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ammonium Hydroxide 6% V/V, 1336-21-6, WT%: <10%, 4l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.