Learn More
Genscript Corporation AmMag™ MR magnetic rack
Catalog No. NC1453939
Encompass
GenScript AmMagTM MR magnetic separation rack enables mid-scale separation of magnetic beads. The rack is designed to separate GenScript’s paramagnetic beads from solutions contained in 15 mL or 50 mL tubes.
Catalog No. NC1453939 Supplier Genscript Corporation Supplier No. L00723
GenScript AmMagTM MR magnetic separation rack enables mid-scale separation of magnetic beads. The rack is designed to separate GenScript’s paramagnetic beads from solutions contained in 15 mL or 50 mL tubes.