Learn More
Genscript Corporation AmMag MR-mini magnetic rack
Catalog No. 5000058065
Encompass
GenScript AmMag sup TM /sup MR-mini magnetic separation rack enables small-scale separation of magnetic beads The rack is designed to separate GenScript s paramagnetic beads from solutions contained in 1 5 mL and 2 mL micro-centrifuge tubes The magnetic field has been optimized for separation of tubes 2 mL in volume
Catalog No. 50-000-58065 Supplier Genscript Corporation Supplier No. L00722
GenScript AmMag sup TM /sup MR-mini magnetic separation rack enables small-scale separation of magnetic beads The rack is designed to separate GenScript s paramagnetic beads from solutions contained in 1 5 mL and 2 mL micro-centrifuge tubes The magnetic field has been optimized for separation of tubes 2 mL in volume