Learn More
Genscript Corporation AmMag Ni Magnetic beads
Supplier:  Genscript Corporation L0077640
Encompass
GenScript s AmMag Ni magnetic beads are designed for purification and screening of histidine-tagged (his-tagged) proteins The beads are highly EDTA and DTT resistant (Table2) enabling them to bind proteins in the presence of chelators The beads are also optimized for easy cleanup and regeneration for reuse with multiple samples Table 1 describes the basic properties of the AmMag Ni magnetic beads
Catalog No. 50-000-57879