Learn More
Genscript Corporation AmMag Protein A Magnetic Beads, 4 ml, alkaline tolerant Protein A, super paramagnetic, 25% (v/v) slurry in 20% ethanol, 40 mg human IgG/ml settled beads
Catalog No. NC2853914
Encompass
AmMag Protein A Magnetic Beads, 4 ml, alkaline tolerant Protein A, super paramagnetic, 25% (v/v) slurry in 20% ethanol, 40 mg human IgG/ml settled beads
Catalog No. NC2853914 Supplier Genscript Corporation Supplier No. L006954
AmMag Protein A Magnetic Beads, 4 ml, alkaline tolerant Protein A, super paramagnetic, 25% (v/v) slurry in 20% ethanol, 40 mg human IgG/ml settled beads