Learn More
Genscript Corporation AmMag Protein A Magnetic Bead
Catalog No. 5000058064
Encompass
GenScript AmMag Protein A Magnetic Beads are developed and optimized for antibody purification for high throughput and high volume applications
Catalog No. 50-000-58064 Supplier Genscript Corporation Supplier No. L0069580
Add to Cart
Add to Cart
Request a Quote
GenScript AmMag Protein A Magnetic Beads are developed and optimized for antibody purification for high throughput and high volume applications