Learn More
Genscript Corporation AmMag Quatro 1100 Automation
Catalog No. 5000058354
Encompass
The AmMag Quatro 1100 automated small-scale magnetic bead purification system is designed to efficiently purify plasmid DNA from Escherichia coli (E coli) using the AmMag Quatro Plasmid Mini kits
Catalog No. 50-000-58354 Supplier Genscript Corporation Supplier No. D00043
Add to Cart
Add to Cart
Request a Quote
The AmMag Quatro 1100 automated small-scale magnetic bead purification system is designed to efficiently purify plasmid DNA from Escherichia coli (E coli) using the AmMag Quatro Plasmid Mini kits