Learn More
Genscript Corporation AmMag Quatro Automation Modul
Catalog No. 5000058945
Encompass
The AmMag Quatro is an automated solution for purification of large-scale high quality transfection-grade plasmids from i Escherichia coli /i cultures
Catalog No. 50-000-58945 Supplier Genscript Corporation Supplier No. D00019
Add to Cart
Add to Cart
Request a Quote
The AmMag Quatro is an automated solution for purification of large-scale high quality transfection-grade plasmids from i Escherichia coli /i cultures