Learn More
Genscript Corporation AmMag Quatro Endofree Plasmid
Catalog No. 5000057866
Encompass
The AmMag Quatro is an automated solution for purification of large-scale high quality transfection-grade plasmids from Escherichia coli cultures The solution consists of a system controller that provides simultaneous operation of up to 4 independent purification modules along with pre-filled consumables and verified protocols
Catalog No. 50-000-57866 Supplier Genscript Corporation Supplier No. L0103124
The AmMag Quatro is an automated solution for purification of large-scale high quality transfection-grade plasmids from Escherichia coli cultures The solution consists of a system controller that provides simultaneous operation of up to 4 independent purification modules along with pre-filled consumables and verified protocols