Learn More
Genscript Corporation AmMag Quatro Plasmid Mini Con
Catalog No. 5000058376
Encompass
The AmMag Quatro Plasmid Mini Consumables 96 Preps contains the 48-Deep Well Plates 4-Column Reagent Reservoir 1000 L Tips Quatro Reaction Plates and 96-Deep Well Plates It is suitable for the AmMag Quatro 1100 Automated small-scale magnetic bead purification system
Catalog No. 50-000-58376 Supplier Genscript Corporation Supplier No. D00056
The AmMag Quatro Plasmid Mini Consumables 96 Preps contains the 48-Deep Well Plates 4-Column Reagent Reservoir 1000 L Tips Quatro Reaction Plates and 96-Deep Well Plates It is suitable for the AmMag Quatro 1100 Automated small-scale magnetic bead purification system