Learn More
Genscript Corporation AmMag Quatro Plasmid Purifica
Catalog No. 5000057867
Encompass
The AmMag Quatro is an automated solution for purification of large-scale high quality transfection-grade plasmids from Escherichia coli cultures The solution consists of a system controller that provides simultaneous operation of up to 4 independent purification modules along with pre-filled consumables and verified protocols
Catalog No. 50-000-57867 Supplier Genscript Corporation Supplier No. L0094324
The AmMag Quatro is an automated solution for purification of large-scale high quality transfection-grade plasmids from Escherichia coli cultures The solution consists of a system controller that provides simultaneous operation of up to 4 independent purification modules along with pre-filled consumables and verified protocols