Learn More
Genscript Corporation AmMag Ultra AT protein A mag
Supplier: Genscript Corporation L0094520
AmMag Ultra AT Protein A Magbeads are developed and optimized for antibody purification for high throughput and high volume applications and can withstand 1M alkali