Learn More
Genscript Corporation AmMag Ultra AT protein A mag
Supplier:  Genscript Corporation L00945200
Encompass
AmMag Ultra AT Protein A Magbeads are developed and optimized for antibody purification for high throughput and high volume applications and can withstand 1M alkali
Catalog No. 50-000-58060
Add to Cart
Request a Quote