Learn More
Genscript Corporation AmMag Ultra AT protein A mag
Catalog No. 5000058401
Encompass
AmMag Ultra AT Protein A Magbeads are developed and optimized for antibody purification for high throughput and high volume applications and can withstand 1M alkali
Catalog No. 50-000-58401 Supplier Genscript Corporation Supplier No. L009454
AmMag Ultra AT Protein A Magbeads are developed and optimized for antibody purification for high throughput and high volume applications and can withstand 1M alkali