Learn More
Genscript Corporation AmMag Wand Sleeve D12
Catalog No. 5000059541
Encompass
GenScript s AmMag Wand enables protein and antibody purification from cultures up to 2 L in volume using the AmMag SA Magnetic beads are directly incubated in cultures or lysates to bind target proteins and antibodies Once binding is complete the AmMag Wand is used to collect the beads and transfer them into 50 ml tubes to be washed and eluted automatically using the AmMag SA purification system or manually using the AmMag MR magnetic racks
Catalog No. 50-000-59541 Supplier Genscript Corporation Supplier No. L00779
GenScript s AmMag Wand enables protein and antibody purification from cultures up to 2 L in volume using the AmMag SA Magnetic beads are directly incubated in cultures or lysates to bind target proteins and antibodies Once binding is complete the AmMag Wand is used to collect the beads and transfer them into 50 ml tubes to be washed and eluted automatically using the AmMag SA purification system or manually using the AmMag MR magnetic racks