Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammelide 98.0+%, TCI America™
SDP

Catalog No. A06455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A06455G 5 g
A06451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A06455G Supplier TCI America Supplier No. A06455G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 645-93-2
Molecular Formula C3H4N4O2
Molecular Weight (g/mol) 128.091
MDL Number MFCD00049046
InChI Key YSKUZVBSHIWEFK-UHFFFAOYSA-N
Synonym 6-Amino-1,3,5-triazine-2,4-diol, Cyanuramide, Cyanuric Acid Monoamide, Dihydro-6-imino-1,3,5-triazine-2,4(1H,3H)-dione, Melanuric Acid
PubChem CID 12584
ChEBI CHEBI:28134
IUPAC Name 6-amino-1H-1,3,5-triazine-2,4-dione
SMILES C1(=NC(=O)NC(=O)N1)N

Specifications

Color White
Quantity 5 g
Formula Weight 128.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ammelide
EINECSNumber (5)-5440
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.