Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammeline 95.0+%, TCI America™
SDP

Catalog No. A067625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A067625G 25 g
A06765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A067625G Supplier TCI America Supplier No. A067625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 645-92-1
Molecular Formula C3H5N5O
Molecular Weight (g/mol) 127.107
MDL Number MFCD00023186
InChI Key MASBWURJQFFLOO-UHFFFAOYSA-N
Synonym ammeline, 4,6-diamino-1,3,5-triazin-2-ol, ammelin, 1,3,5-triazin-2 1h-one, 4,6-diamino, 2,4-diamino-6-hydroxy-1,3,5-triazine, cyanurodiamide, 2,4-diamino-1,3,5-triazin-6-one, 4,6-diamino-1,3,5-triazin-2 1h-one, unii-lc8g9556q0, s-triazin-2-ol, 4,6-diamino
PubChem CID 12583
ChEBI CHEBI:28646
IUPAC Name 2,6-diamino-1H-1,3,5-triazin-4-one
SMILES C1(=NC(=O)N=C(N1)N)N

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 127.11
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Ammeline
EINECSNumber (5)-5439
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.