Learn More
Enterprise Technology Solutions AMMEX Exam Gloves,Nitrile,Powder-Free,Latex-Free,Grip,Chemo-Rated,3Mil,Large,100/BX,Cerulean
SDP
Catalog No. 5000828382
Encompass_Preferred
AMMEX Professional Exam Blue Nitrile Gloves provide reliable protection for healthcare professionals. Designed for durability and comfort, these gloves are chemo-rated and resist fentanyl permeation for up to 240 minutes. They offer consistent barrier protection against bodily fluids, bloodborne pathogens and contaminants to ensure safety in demanding environments. Powder-free and latex-free, they deliver superior grip and tactile sensitivity for precision tasks.
Catalog No. 50-008-28382 Supplier Enterprise Technology Solutions Supplier No. AMXACNPF46100
AMMEX Professional Exam Blue Nitrile Gloves provide reliable protection for healthcare professionals. Designed for durability and comfort, these gloves are chemo-rated and resist fentanyl permeation for up to 240 minutes. They offer consistent barrier protection against bodily fluids, bloodborne pathogens and contaminants to ensure safety in demanding environments. Powder-free and latex-free, they deliver superior grip and tactile sensitivity for precision tasks.