Learn More
Quantimetrix Ammonia/ Alcohol Control, Liquid, ready-to-use. Three-year refrigerated shelf life and 4-month open vial stability when stored at 2-8°C. Each level sold separately. Mid, Level 2, 3x5mL. 57-55-6
SDP
Catalog No. 501528624
Encompass_Preferred
Ammonia/ Alcohol Control, Liquid, ready-to-use. Three-year refrigerated shelf life and 4-month open vial stability when stored at 2-8°C. Each level sold separately. Mid, Level 2, 3x5mL. 57-55-6
This item is not returnable. View return policy
Catalog No. 50-152-8624 Supplier Quantimetrix Supplier No. 131231
Ammonia/ Alcohol Control, Liquid, ready-to-use. Three-year refrigerated shelf life and 4-month open vial stability when stored at 2-8°C. Each level sold separately. Mid, Level 2, 3x5mL. 57-55-6