Learn More
Quantimetrix Ammonia/ Alcohol Control, Liquid, ready-to-use. Three-year refrigerated shelf life and 4-month open vial stability when stored at 2-8°C. Each level sold separately. High, Level 3, 3x5mL. 57-55-6
SDP
Catalog No. 501528625
Encompass_Preferred
Ammonia/ Alcohol Control, Liquid, ready-to-use. Three-year refrigerated shelf life and 4-month open vial stability when stored at 2-8°C. Each level sold separately. High, Level 3, 3x5mL. 57-55-6
This item is not returnable. View return policy
Catalog No. 50-152-8625 Supplier Quantimetrix Supplier No. 131331
Ammonia/ Alcohol Control, Liquid, ready-to-use. Three-year refrigerated shelf life and 4-month open vial stability when stored at 2-8°C. Each level sold separately. High, Level 3, 3x5mL. 57-55-6