Learn More
ERA WATERS CORP AMMONIA AS NITROGEN STD 500ML
SDP
Catalog No. NC9704312
Encompass
Ammonia as Nitrogen (N) - 1000mg/L, 500mL, Designed to be used either as calibration or calibration-check standards, Traceable to NIST Standard Reference Materials where possible, Now includes manufacturing uncertainties, Analyte: 1000mg/L
Catalog No. NC9704312 Supplier ERA WATERS CORP Supplier No. 985
Ammonia as Nitrogen (N) - 1000mg/L, 500mL, Designed to be used either as calibration or calibration-check standards, Traceable to NIST Standard Reference Materials where possible, Now includes manufacturing uncertainties, Analyte: 1000mg/L