Learn More
Cederlane Ammonia Assay Kit Rapid, 96 assays (manual) / 960 assays (microplate) / 960 assays (auto-analyser), Absorbance detection at 340 nm, Reaction time 5 min
SDP
Catalog No. NC1610867
Encompass
The CEDARLANE Ammonia Assay Kit (Rapid) is a rapid enzymatic assay used for measuring ammonia across various sample types, including grape juice and food products. Utilizing uninhibited glutamate dehydrogenase, this kit provides fast and accurate results, making it ideal for quality control and research applications.
Catalog No. NC1610867 Supplier Cederlane Supplier No. 700004261
The CEDARLANE Ammonia Assay Kit (Rapid) is a rapid enzymatic assay used for measuring ammonia across various sample types, including grape juice and food products. Utilizing uninhibited glutamate dehydrogenase, this kit provides fast and accurate results, making it ideal for quality control and research applications.