Learn More
CHEMTEQ INC AMMONIA BREAKTH-INDIC STCK 5PK
Catalog No. NC2456231
Encompass
Dimensions: Diameter = 38.1mm (1.5in), Height = 2.0mm (0.11in)Shelf life: 2 years at 4oC to 25oC, (39oF to 77oF)Service life: 4 months
Catalog No. NC2456231 Supplier CHEMTEQ INC Supplier No. 251
Dimensions: Diameter = 38.1mm (1.5in), Height = 2.0mm (0.11in)Shelf life: 2 years at 4oC to 25oC, (39oF to 77oF)Service life: 4 months