Learn More
Hach Company Ammonia Buffer, EDTA/NaOH Solution, 1 L
Catalog No. NC1952465 Shop All Hach Company Products
Encompass
Ammonia Buffer, EDTA/NaOH Solution is a ready-to-use reagent supplied in a 1 L bottle for analytical determination of ammonia by flow injection analysis (FIA). The formulation contains EDTA and sodium hydroxide to condition samples and improve accuracy for standard FIA ammonia methods. Follow manufacturer storage and handling instructions and consult the Safety Data Sheet (SDS) before use.
  • Ready-to-use 1 L reagent bottle.
  • Formulated with EDTA and sodium hydroxide to buffer samples.
  • Intended for ammonia analysis by flow injection analysis (FIA).
  • Suitable for use with unpreserved samples in standard ammonia methods.
  • Safety Data Sheet available; follow storage and handling instructions.
Catalog No. NC1952465 Supplier Hach Company Supplier No. 52017
Ammonia Buffer, EDTA/NaOH Solution is a ready-to-use reagent supplied in a 1 L bottle for analytical determination of ammonia by flow injection analysis (FIA). The formulation contains EDTA and sodium hydroxide to condition samples and improve accuracy for standard FIA ammonia methods. Follow manufacturer storage and handling instructions and consult the Safety Data Sheet (SDS) before use.
  • Ready-to-use 1 L reagent bottle.
  • Formulated with EDTA and sodium hydroxide to buffer samples.
  • Intended for ammonia analysis by flow injection analysis (FIA).
  • Suitable for use with unpreserved samples in standard ammonia methods.
  • Safety Data Sheet available; follow storage and handling instructions.