Learn More
Hach Company Ammonia Cyanurate Reagent Powder Pillows, 10 mL, pk/100
Supplier:  Hach Company 2653199
Encompass
For determination of Ammonia Nitrogen by the Salicylate method. Hach Method 8155. Range: 0.01 - 0.50 mg/L NH3-N. Sample Size: 10 mL. Pack of 100 Powder Pillows.
Catalog No. 50-199-6439