Learn More
MSC AMMONIA DETECTOR TUBE
Catalog No. 5000792779
Encompass
3LA 2.5-200 PPM 10/PK AMMONIA DETECTOR TUBE
Catalog No. 50-007-92779 Supplier MSC Supplier No. 54508650
3LA 2.5-200 PPM 10/PK AMMONIA DETECTOR TUBE