Learn More
Perkin Elmer US LLC Ammonia Dual-Stage High-Purity Regulator
Supplier:  Perkin Elmer US LLC N8152490
Encompass_Preferred
Benefits and Features: Stainless steel barstock body Stainless steel diaphragm Metal to metal seals minimize diffusion of contaminants 2-1/2 inlet and delivery pressure gauges Equipped with outlet diaphragm valve Bonnet ported and threaded to pipe gases away from work area 1/4 compression tube outlet connection 10 micron metal filter protects seat from contamination 1/8 Ultra Torr Fitting
Catalog No. 50-210-3927