Learn More
Hach Company Conditioning Solution, ISENH3181, 500 mL
Catalog No. 501996197 Shop All Hach Company Products
Encompass
Used as a conditioning solution between samples Conditioning solution for Ammonia Ion Selective Electrode (ISENH3181), 500 mL bottleThis solution is for use between sample tests. It will help to reset the internal pH, and provide quicker, more accurate readings.(This solution is not to be used for overnight storage, or storage over one week, please see ISENH3181 electrode manual.)
Catalog No. 50-199-6197 Supplier Hach Company Supplier No. 2541249
Used as a conditioning solution between samples Conditioning solution for Ammonia Ion Selective Electrode (ISENH3181), 500 mL bottleThis solution is for use between sample tests. It will help to reset the internal pH, and provide quicker, more accurate readings.(This solution is not to be used for overnight storage, or storage over one week, please see ISENH3181 electrode manual.)