Learn More
Hach Company Filling Solution, ISENH3181, 0.1 M NH₄Cl, 50 mL
Supplier:  Hach Company 4447226
Encompass
Filling solution for Ammonia Ion Selective Electrode (ISENH3181), Ammonium Chloride (NH₄Cl), 0.1 M, 50 mL bottle
Catalog No. 50-199-7035