Learn More
HANNA INSTRUMENTS INC Ammonia High Range Reagents with Barcode Recognition
SDP
Catalog No. NC1660478
Encompass
The HI94764B-25 are reagents that follow an adaptation of the ASTM Manual of Water and Environmental Technology, D1426-92, Nessler method for the determination of high range ammonia using the compatible HI83224 benchtop photometer. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each box for traceability.Pre-dosed reagents for ease of useSupplied with certificate of qualityMarked with expiration date and lot number for traceability
Catalog No. NC1660478 Supplier HANNA INSTRUMENTS INC Supplier No. HI94764B25
The HI94764B-25 are reagents that follow an adaptation of the ASTM Manual of Water and Environmental Technology, D1426-92, Nessler method for the determination of high range ammonia using the compatible HI83224 benchtop photometer. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each box for traceability.Pre-dosed reagents for ease of useSupplied with certificate of qualityMarked with expiration date and lot number for traceability