Learn More
HANNA INSTRUMENTS INC Ammonia High Range Reagents (100 tests)
SDP
Catalog No. NC1890595
Encompass
The HI93733-01 are reagents for the colorimetric determination of ammonium (NH4+). There are enough reagents for 100 tests to be used with Hanna's compatible portable photometer. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each bottle for traceability.Pre-made reagents for ease of usePrepared with high purity chemicalsMarked with expiration date and lot number for traceability
Catalog No. NC1890595 Supplier HANNA INSTRUMENTS INC Supplier No. HI9373301
The HI93733-01 are reagents for the colorimetric determination of ammonium (NH4+). There are enough reagents for 100 tests to be used with Hanna's compatible portable photometer. These high quality reagents are manufactured in our state-of-the-art facility and are clearly marked with the lot number and expiration date on each bottle for traceability.Pre-made reagents for ease of usePrepared with high purity chemicalsMarked with expiration date and lot number for traceability