Learn More
Hach Company Ammonia Ionic Strength Adjustor (ISA) Powder Pillows, pk/100
Catalog No. 501997034 Shop All Hach Company Products
Encompass
Ionic Strength Adjustor (ISA) for Ammonia determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Ammonia level readings. Eliminate most interferences. Pack of 100 powder pillows.
Exact match for part number 4447169
Catalog No. 50-199-7034 Supplier Hach Company Supplier No. 4447169
Ionic Strength Adjustor (ISA) for Ammonia determinations by Ion Selective Electrode (ISE) method. Powder form prevent from diluting samples. Ideal for low Ammonia level readings. Eliminate most interferences. Pack of 100 powder pillows.
Exact match for part number 4447169