Learn More
Hach Company Ammonia Ionic Strength Adjustor (ISA), Solution, 500 mL
Supplier:  Hach Company 2824349
Encompass
Ionic Strength Adjustor (ISA) for Ammonia determinations by Ion Selective Electrode (ISE) method. Eliminate most interferences. Solution, 500 mL bottle.
Catalog No. 50-199-6724
 Restricted Item