Learn More
CHEMetrics™ by AquaPhoenix Ammonia/Monochloramine Refill Kit



Description
- Free ammonia reacts with hypochlorite to form monochloramine. Monochloramine reacts with hydroxybenzyl alcohol (HBA), in the presence of sodium nitroferricyanide, to form a green-colored complex. This method measures the sum of free ammonia and monochloramine.
- Results are expressed as ppm (mg/L) ammonia-nitrogen (NH3-N). To convert results to ppm ammonia (NH3), multiply by 1.22.
- To differentiate between monochloramine and free ammonia:
- Determine the monochloramine + free ammonia concentration (as ppm NH3-N): Analyze the sample according to the kit instructions.
- Determine the monochloramine concentration (as ppm NH3-N): Analyze the sample, omitting the addition of A1406 Activator Solution (hypochlorite). Note: To convert results from ppm NH3-N to ppm monochloramine as chlorine (NH2Cl-Cl2), multiply by 5.
- Determine the free ammonia concentration (as ppm NH3-N): Subtract the result obtained in Step 2 (in ppm NH3-N) from the result obtained in Step 1.
Specifications
Specifications
| Analytes | Ammonia/Monochloramine |
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-1420 Ammonia or K-6802 Monochloramine Test Kits |
| Quantity | 30/Pk. |
| Sterility | Non-sterile |
| Type | Ammonia/Monochloramine Refill Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.