Learn More
Hach Company Intellical™ ISENH3181 Ammonia (NH₃) Ion Selective Electrode (ISE), 3 m cable
Catalog No. 501997728 Shop All Hach Company Products
Encompass
Intellical™ ISENH3181 is a digital, combination gas-sensing, ammonia ion selective electrode (ISE) with a with a replaceable membrane module, refillable outer body, a non-refillable gel 3M KCl reference with a double junction porous PTFE annular ring and built-in temperature sensor. The electrode measures ammonia concentration in water samples. A 50 mL bottle of 0,1 M NH4Cl Electrode Filling Solution is included with the probe.The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISENH3181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENH3181 is ideal for measuring ammonia concentrations in wastewater, drinking water and general water quality applications.
Catalog No. 50-199-7728 Supplier Hach Company Supplier No. ISENH318103
Add to Cart
Add to Cart
Intellical™ ISENH3181 is a digital, combination gas-sensing, ammonia ion selective electrode (ISE) with a with a replaceable membrane module, refillable outer body, a non-refillable gel 3M KCl reference with a double junction porous PTFE annular ring and built-in temperature sensor. The electrode measures ammonia concentration in water samples. A 50 mL bottle of 0,1 M NH4Cl Electrode Filling Solution is included with the probe.The laboratory version of this ISE is shockproff with its Epoxy plastic body.The Intellical ISENH3181 probe is available with a 1 or 3 meter cable and is intended for laboratory use. The ISENH3181 is ideal for measuring ammonia concentrations in wastewater, drinking water and general water quality applications.