Learn More
Hach Company Ammonia Salicylate Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501996440 Shop All Hach Company Products
Encompass
For determination of Ammonia Nitrogen by the Salicylate method. Hach Method 8155. Range: 0.01 - 0.50 mg/L NH3-N. Sample Size: 10 mL. Pack of 100 Powder Pillows.
Catalog No. 50-199-6440 Supplier Hach Company Supplier No. 2653299
For determination of Ammonia Nitrogen by the Salicylate method. Hach Method 8155. Range: 0.01 - 0.50 mg/L NH3-N. Sample Size: 10 mL. Pack of 100 Powder Pillows.