Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonia Solution 28-30%, Supelco

Catalog No. 11100260 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-260 1 L
11-100-254 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-260 Supplier MilliporeSigma Supelco Supplier No. 1054231011
 Restricted Item
Add to Cart
Add to Cart

For analysis EMSURE™ ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonia Solution 28-30%
CAS 1336-21-6
Quantity 1 L
Molecular Weight (g/mol) 35.05
Percent Purity Unknown
Grade ACS
Packaging HDPE Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.