Learn More
CHEMetrics™ by AquaPhoenix Ammonia Stabilizer Solution Pack, Use with K-1513 and K-1510
SDP
Catalog No. 13299554
Change view
Click to view available options
Quantity:
6 x 20 mL Bottles
Catalog No. Quantity
13-299-554 6 x 20 mL Bottles
1 options
Catalog No. 13-299-554 Supplier CHEMetrics™ by AquaPhoenix Supplier No. A1501
Add to Cart
Add to Cart
Ammonia stabilizer solution for use with K-1513 and K-1510 test kits. Designed for seawater analysis applications.
  • Provides sufficient reagent for approximately 100 seawater analyses
Specifications
pH Range 7
Volume (Metric) 120 mL
Quantity 6 x 20 mL Bottles
Type Ammonia Stabilizer Solution