Learn More
Aqua Solutions Nitrogen Standard 1,000 ppm Solution,(1ml = 1mg N) Certified (250ML)
SDP
Catalog No. NC0951433
Encompass_Preferred
Nitrogen Standard 1,000 ppm Solution,(1ml = 1mg N) Certified (250ML)
Catalog No. NC0951433 Supplier Aqua Solutions Supplier No. 6490250ML
Nitrogen Standard 1,000 ppm Solution,(1ml = 1mg N) Certified (250ML)