Learn More
YSI 3P LifeScience AMMONIA STANDARD 500MG/L 250ML
Catalog No. NC0493992
Encompass
Ammonium Standard, 500 Mg/L (250 Ml). 500 Mg/L Ammonium Standard Solution For Use With Ysi 2950 Analyzer. Default Calibrator Value For The 2950 Analyzer.
Catalog No. NC0493992 Supplier YSI 3P LifeScience Supplier No. 2972
Ammonium Standard, 500 Mg/L (250 Ml). 500 Mg/L Ammonium Standard Solution For Use With Ysi 2950 Analyzer. Default Calibrator Value For The 2950 Analyzer.